4'-Bromo-2'-fluoroacetophenone 96%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-(4-Bromo-2-fluorophenyl)ethanone
H62127.03 H62127.06
H62127.03EA 4175 SEK
H62127.03 H62127.06
4'-Bromo-2'-fluoroacetophenone 96%
4'-Bromo-2'-fluoroacetophenone
Purity: 96% CAS No.: 625446-22-2 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Physical Form: Solid
Formel: C₈H₆BrFO
MW: 217.04 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03411551
CAS nummer: 625446-22-2