Salicylaldehyde azine ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Salicylaldazine

S0850-5G S0850-25G
TCIAS0850-5GEA 285 SEK
TCIAS0850-5G TCIAS0850-25G
Salicylaldehyde azine ≥98.0%
Salicylaldehyde azine
Formel: C₁₄H₁₂N₂O₂
MW: 240,26 g/mol
MDL Number: MFCD00043496
CAS nummer: 959-36-4
EINECS: 213-500-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea4900a21732c40dd9256621d98b12aa092a73f6&vid=ea4900a21732c40dd9256621d98b12aa092a73f6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 216.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR