Kalciumklorid dihydrat, EMPROVE® API BP, Ph. Eur., JP, USP suitable for use as an active pharmaceutical ingredient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Varning
Synonyms: Calcium Chloride Dihydrate, Calcium dichloride
1.02465.1000 1.02465.9029 1.02465.9525
1.02465.1000EA 2625 SEK
1.02465.1000 1.02465.9029 1.02465.9525
Kalciumklorid dihydrat, EMPROVE® API BP, Ph. Eur., JP, USP suitable for use as an active pharmaceutical ingredient, SAFC®
Kalciumklorid dihydrat
Formel: CaCl₂·2H₂O
MW: 147.01 g/mol
Kokpunkt: 1600
Smältpunkt: 176
Densitet: 1.85 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149613
CAS nummer: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660