Drum sumps, large volumes, GW
Leverantör: LACONT UMWELTTECHNIK
P21-1103-A P21-1104-A P21-1105-A
LACOP21-1103-AEA 0 SEK
LACOP21-1103-A LACOP21-1104-A LACOP21-1105-A
Drum sumps, large volumes, GW
Förvaring av tunnor
These drum sumps made of 3 mm fire galvanised sheet steel are used for storing and filling off water-endangering and flammable liquids.
  • Removable fire-galvanised grid
  • Ground clearance for fork-lift transport
  • Models GW 3, GW 5 and GW 6 suitable for storing 1000 L containers
Leveransinformation: Models GW 4, GW 5 and GW 6 are supplied as single sump pallets, the single sump pallet are connected together on site with sump pallet connectors to form a single storage surface.