Paraffin, Solidification point: 54 - 56 °C, Q Path® Microscopy
Leverantör: VWR Q-Path Chemicals
Synonyms: Wax Paraffin, Paraffin wax
10048502.EA 1750 SEK
10048502. 10048500. 10048501.
Paraffin, Solidification point: 54 - 56 °C, Q Path® Microscopy
Paraffin
Wax (54 - 56 °C) for embedding and tissue processing of histological samples. This Paraffin is DMSO free.
Smältpunkt: 54…56 °C
CAS nummer: 8002-74-2
EINECS: 232-315-6
IVDLogo