Papanicolaou lösning EA 50, Q Path® för mikroskopi
Leverantör: VWR Q-Path Chemicals
Fara
10047011.EA 1975 SEK
10047011. 10047111. 00607141.
Papanicolaou lösning EA 50, Q Path® för mikroskopi
Papanicolaou's solution EA 50
Ready to use solution for Papanicolaou cytological staining
IVD registered. Instructions for use on vwr.com- just search for the product.
Densitet: 0,82 g/cm³ (20 °C)
Flampunkt: 17 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00147831
UN: 1992
ADR: 3 (6.1),II
IVDLogo
Order Now
Specifikation testresultat
Appearance Clear liquid
Cyanophilic cytoplasm Blue-green
Eosinophilic cytoplasm Pink
Nuclei Blue to dark violet
Suitable for microscopy (vaginal smear) Passes test