Fenol kondenserad ≥99.0% mättad lösning i vatten pH 6.6
Leverantör: AMBION
Fara
9712
AMBI9712EA 0 SEK
AMBI9712
Fenol kondenserad ≥99.0% mättad lösning i vatten pH 6.6
Fenol kondenserad
Ambion® Water Saturated Phenol is supplied premixed and at pH 6.6 (pH 7.9 ± 0.2, with included buffer). Water Saturated Phenol is used in RNA and DNA extraction procedures. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion® premixed, quality tested, saturated phenols are ready-to-use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.
Formel: C₆H₅OH
MW: 94,11 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00002143
CAS nummer: 108-95-2
EINECS: 203-632-7
UN: 2821
ADR: 6.1,II