D-Glukonsyrans-1 ,5-lakton ≥98.0%

Leverantör: TCI EUROPE

Synonyms: GDL, D-Glucono-δ-lactone, D-Gluconic acid-1,5-lactone, delta-Gluconolactone, D-Gluconic acid lactone, (3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, D-(+)-Glucono-δ-lactone, D-(+)-Gluconic acid δ-lactone, D-Gluconic acid-δ-lacton, D-(+)-Glucono-1,5-lactone, Gluconolactone

G0039-25G G0039-500G
TCIAG0039-25GEA 230 SEK
TCIAG0039-25G TCIAG0039-500G
D-Glukonsyrans-1 ,5-lakton ≥98.0%
D-Glukonsyrans-1 ,5-lakton
Formel: C₆H₁₀O₆
MW: 178,14 g/mol
Kokpunkt: 385 °C (1013 hPa)
Smältpunkt: 153 °C
Densitet: 1,61 g/cm³ (-5 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006647
CAS nummer: 90-80-2
EINECS: 202-016-5
Merck Index: 14,04457
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c71f1736ada2f0dbb12feee0d74a4d066142a0b6&vid=c71f1736ada2f0dbb12feee0d74a4d066142a0b6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization back titration) min. 98.0 %
Specific rotation [a]20/D +60.0 to +67.0 deg (C=10, H2O)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR