4-Chloro-2-picoline ≥98.0%

Leverantör: TCI
Varning

M2284-1G M2284-5G
TCIAM2284-1GEA 230 SEK
TCIAM2284-1G TCIAM2284-5G
4-Chloro-2-picoline ≥98.0%
4-Chloro-2-picoline
Formel: C₆H₆ClN
Kokpunkt: 56°C
MDL Number: MFCD00038833
CAS nummer: 3678-63-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2cb52fd6be7fdeb626aae786bc340a9121bc275&vid=d2cb52fd6be7fdeb626aae786bc340a9121bc275&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR