2-Bromo-3-methoxynaphthalene ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
B3403-1G
B3403-5G
TCIAB3403-1GEA
805
SEK
TCIAB3403-1G
TCIAB3403-5G
2-Bromo-3-methoxynaphthalene ≥97.0%
2-Bromo-3-methoxynaphthalene
Formel:
C₁₁H₉BrO MW: 237,0926 g/mol |
MDL Number:
MFCD11870110 CAS nummer: 68251-77-4 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 73.0 to 78.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...