Trimellitic acid trimethyl ester ≥95.0%
Leverantör: TCI EUROPE
Synonyms: Trimethyl 1,2,4-benzenetricarboxylate
T1229-25G T1229-500G
TCIAT1229-25GEA 215 SEK
TCIAT1229-25G TCIAT1229-500G
Trimellitic acid trimethyl ester ≥95.0%
Trimellitic acid trimethyl ester
Formel: C₁₂H₁₂O₆
MW: 252,22 g/mol
Kokpunkt: 189°C
Flampunkt: 182°C
MDL Number: MFCD00043629
CAS nummer: 2459-10-1
EINECS: 219-547-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22d11f06f875a526fa1e2029e20984d9c8a54783&vid=22d11f06f875a526fa1e2029e20984d9c8a54783&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %