3-Amino-2-chloro-6-methylpyridine 98+%
Leverantör: Thermo Fisher Scientific
|
Varning
|
360740010
360740050
ACRO360740010EA
1225
SEK
ACRO360740010
ACRO360740050
3-Amino-2-chloro-6-methylpyridine 98+%
3-Amino-2-chloro-6-methylpyridine
Formel:
C₆H₇ClN₂ Smältpunkt: 74…80 °C Storage Temperature: Omgivande |
MDL Number:
MFCD03095213 CAS nummer: 39745-40-9 UN: 2811 ADR: 6.1,II |
Specifikation testresultat
Appearance | Cream to tan Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥98.0 % |