2-Amino-3,5-dichloro-4-methylpyridine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 3,5-Dichloro-4-methylpyridin-2-amine

A1932-1G A1932-5G
TCIAA1932-1GEA 455 SEK
TCIAA1932-1G TCIAA1932-5G
2-Amino-3,5-dichloro-4-methylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-3,5-dichloro-4-methylpyridine
Formel: C₆H₆Cl₂N₂
MW: 177,0300 g/mol
MDL Number: MFCD00129031
CAS nummer: 31430-47-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e15291792826f6ff08ee8b238a70110fd12d51f2&vid=e15291792826f6ff08ee8b238a70110fd12d51f2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 123.0 to 127.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR