2,2,4-Trimethyl-1,3-dioxolane ≥99%
Leverantör: Thermo Fisher Scientific
Varning
B21823.14 B21823.22 B21823.36
B21823.14EA 1050 SEK
B21823.14 B21823.22 B21823.36
2,2,4-Trimethyl-1,3-dioxolane ≥99%
2,2,4-Trimethyl-1,3-dioxolane
Purity: 99% CAS No.: 1193-11-9 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 32°(90°F) Boiling Point: 98° UN No.: 3271 MDL No.: MFCD00090841
Formel: C₆H₁₂O₂
Kokpunkt: 98 °C
Densitet: 0,897
Flampunkt: 32 °C (90 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00090841
CAS nummer: 1193-11-9
EINECS: 214-766-0
ADR: 3,III