2-Methoxy-4-methylpyridine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
M2190-1G
M2190-5G
TCIAM2190-1GEA
445
SEK
TCIAM2190-1G
TCIAM2190-5G
2-Methoxy-4-methylpyridine ≥98.0%
2-Methoxy-4-methylpyridine
Formel:
C₇H₉NO MW: 123,1525 g/mol Kokpunkt: 50°C Flampunkt: 53°C |
MDL Number:
MFCD06200799 CAS nummer: 100848-70-2 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...