Direct red 80

Synonyms: CI 35780, Sirius Red F3B, Sirius Red, Picro-Sirius Red

1A-280.0010 1A-280.0025 1A-280.0100
CHRO1A-280.0010EA 58.28 SEK
CHRO1A-280.0010 CHRO1A-280.0025 CHRO1A-280.0100
Direct red 80
Direct red 80
Formel: C₄₅H₂₆N₁₀Na₆O₂₁S₆
MW: 1373,051 g/mol
MDL Number: MFCD00054389
CAS nummer: 2610-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR