2-Acetoxybenzonitrile ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Cyanophenyl acetate

A1118-25G A1118-5G
TCIAA1118-25GEA 1225 SEK
TCIAA1118-25G TCIAA1118-5G
2-Acetoxybenzonitrile ≥97.0%
2-Acetoxybenzonitrile
Formel: C₉H₇NO₂
MW: 161,16 g/mol
Kokpunkt: 254°C
MDL Number: MFCD00017360
CAS nummer: 5715-02-6
EINECS: 227-210-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d05cdc2a79e5604e8ceaa432c27984b57f7d201&vid=1d05cdc2a79e5604e8ceaa432c27984b57f7d201&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR