Sodium tetraethylborate 20% THF
Leverantör: LGC Standards PROMOCHEM
DE-MS-03402S-5 DEMS03402S5X25 DE-MS03402S-25
PROODE-MS-03402S-5EA 0 SEK
PROODE-MS-03402S-5 PROODEMS03402S5X25 PROODE-MS03402S-25
Sodium tetraethylborate 20% THF
Sodium tetraethylborate
Formel: C₈H₂₀BNa
MW: 150.05 g/mol
MDL Number: MFCD00061547
CAS nummer: 15523-24-7