3-(Pentylthio)-1H-1,2,4-triazole

Leverantör: Thermo Scientific

RJC03578DA RJC03578EA
MAYBRJC03578DAEA 0 SEK
MAYBRJC03578DA MAYBRJC03578EA
3-(Pentylthio)-1H-1,2,4-triazole
3-(Pentylthio)-1H-1,2,4-triazole
Formel: C₇H₁₃N₃S
CAS nummer: 71705-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR