1-(2-Cyanophenyl)piperidine ≥97%
Leverantör: Thermo Fisher Scientific
Synonyms: 2-(1-Piperidinyl)benzonitrile, 2-Piperidinobenzonitrile
A11784.03 A11784.06
A11784.03EA 325 SEK
A11784.03 A11784.06
1-(2-Cyanophenyl)piperidine ≥97%
1-(2-Cyanophenyl)piperidine
Purity: 97% CAS No.: 72752-52-4 Molecular Formula: C12H14N2 Formula Weight: 186.26 Melting Point: 41-45° Boiling Point: 122-125°/1mm UN No.: 3077 MDL No.: MFCD00049221
Formel: C₁₂H₁₄N₂
MW: 186.26 g/mol
Kokpunkt: 122…125 °C (1 mmHg)
Smältpunkt: 41…45 °C
Storage Temperature: Omgivande
MDL Number: MFCD00049221
CAS nummer: 72752-52-4
EINECS: 427-330-1
ADR: 9 III