2-(Tributylstannyl)thiazole 96%
Leverantör: Thermo Scientific
Fara
H51090.03 H51090.06
H51090.03EA 3650 SEK
H51090.03 H51090.06
2-(Tributylstannyl)thiazole 96%
2-(Tributylstannyl)thiazole
Purity: 96% CAS No.: 121359-48-6 Molecular Formula: C15H29NSSn Formula Weight: 374.17 Boiling Point: 307-309° UN No.: 2788 MDL No.: MFCD01319057
Formel: C₁₅H₂₉NSSn
MW: 374,17 g/mol
Kokpunkt: 307…309 °C
Densitet: 1,19
Storage Temperature: Omgivande
MDL Number: MFCD01319057
CAS nummer: 121359-48-6
ADR: 6.1,III