2-Ethynylthiophene ≥96.0%
Leverantör: TCI EUROPE
|
Varning
|
E0915-5G
E0915-1G
TCIAE0915-5GEA
2875
SEK
TCIAE0915-5G
TCIAE0915-1G
2-Ethynylthiophene ≥96.0%
2-Ethynylthiophene
Formel:
C₆H₄S MW: 108,1610 g/mol Kokpunkt: 46°C Storage Temperature: Kylskåp |
MDL Number:
MFCD03425129 CAS nummer: 4298-52-6 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Yellow to Amber to Dark purple clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...