Iodotrimethylsilane ≥95.0% stabilised

Leverantör: TCI
Fara

Synonyms: Trimethyliodosilane, Trimethylsilyl iodide

Stabilization: Stabilised with aluminium

I0308-25G I0308-5G
TCIAI0308-25GEA 615 SEK
TCIAI0308-25G TCIAI0308-5G
Iodotrimethylsilane ≥95.0% stabilised
Iodotrimethylsilane
Formel: C₃H₉ISi
Kokpunkt: 106 °C (1013 hPa)
Smältpunkt: < 0 °C
Densitet: 1,47 g/cm³ (20 °C)
Flampunkt: –31°C
MDL Number: MFCD00001028
CAS nummer: 16029-98-4
EINECS: 240-171-0
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5795c365bcf0b20351542c3bd16860bfaa4c2bd&vid=e5795c365bcf0b20351542c3bd16860bfaa4c2bd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Brown clear liquid
Purity (Potassium iodate Method) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR