Methyl-4-amino-3-chlorobenzoate 99%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 4-amino-3-chlorobenzoate
B22519.06 B22519.03
B22519.06EA 3450 SEK
B22519.06 B22519.03
Methyl-4-amino-3-chlorobenzoate 99%
Methyl-4-amino-3-chlorobenzoate
Purity: 99% CAS No.: 84228-44-4 Molecular Formula: C8H8ClNO2 Formula Weight: 185.61 Melting Point: 112-113° MDL No.: MFCD01320686
Formel: C₈H₈ClNO₂
MW: 185,61 g/mol
Smältpunkt: 112…113 °C
Storage Temperature: Omgivande
MDL Number: MFCD01320686
CAS nummer: 84228-44-4