1-(2-Aminoethyl)piperidine ≥97.0% (by GC, titration analysis)
Leverantör: TCI
|
Fara
|
Synonyms:
N-(2-Aminoethyl)piperidine
A1026-25ML
A1026-5ML
TCIAA1026-25MLEA
1275
SEK
TCIAA1026-25ML
TCIAA1026-5ML
1-(2-Aminoethyl)piperidine ≥97.0% (by GC, titration analysis)
1-(2-Aminoethyl)piperidine
Formel:
C₇H₁₆N₂ Kokpunkt: 184°C Flampunkt: 58°C |
MDL Number:
MFCD00006516 CAS nummer: 27578-60-5 EINECS: 248-540-8 UN: 2734 ADR: 8,II |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...