4-Methoxy-3-nitrobenzonitrile

Leverantör: Thermo Scientific

Synonyms: 3-Nitro-4-methoxybenzonitrile

SPB07277DA SPB07277EA
MAYBSPB07277DAEA 0 SEK
MAYBSPB07277DA MAYBSPB07277EA
4-Methoxy-3-nitrobenzonitrile
4-Methoxy-3-nitrobenzonitrile
Formel: C₈H₆N₂O₃
MW: 178.1448 g/mol
MDL Number: MFCD00221439
CAS nummer: 33224-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR