Methyl-2-chloroisonicotinate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Methyl 2-chloropyridine-4-carboxylate, Methyl 2-chloroisonicotinate

M2164-25G M2164-5G
TCIAM2164-25GEA 1700 SEK
TCIAM2164-25G TCIAM2164-5G
Methyl-2-chloroisonicotinate ≥98.0%
Methyl-2-chloroisonicotinate
Formel: C₇H₆ClNO₂
MW: 171,5810 g/mol
Kokpunkt: 70°C
Storage Temperature: Kylskåp
MDL Number: MFCD01765409
CAS nummer: 58481-11-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b6b13c8ac902301fb0ad0d85f8ec0cbc53d4411&vid=0b6b13c8ac902301fb0ad0d85f8ec0cbc53d4411&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Melting point 32.0 to 36.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR