8-Methoxyquinaldine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
8-Methoxy-2-methylquinoline
M1088-1G
M1088-5G
TCIAM1088-1GEA
1075
SEK
TCIAM1088-1G
TCIAM1088-5G
8-Methoxyquinaldine ≥98.0% (by GC, titration analysis)
8-Methoxyquinaldine
Formel:
C₁₁H₁₁NO MW: 173,2111 g/mol |
MDL Number:
MFCD00034436 CAS nummer: 3033-80-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...