1-Bromo-4-dodecylbenzene ≥90.0%

Leverantör: TCI EUROPE

B1612-25G B1612-5G
TCIAB1612-25GEA 3825 SEK
TCIAB1612-25G TCIAB1612-5G
1-Bromo-4-dodecylbenzene ≥90.0%
1-Bromo-4-dodecylbenzene
Formel: C₁₈H₂₉Br
MW: 325,33 g/mol
Kokpunkt: 191°C
MDL Number: MFCD00191386
CAS nummer: 126930-72-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04a3bcba9c666c78ae5b4b11bd46a5bab8f6b4bc&vid=04a3bcba9c666c78ae5b4b11bd46a5bab8f6b4bc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR