Methyl trifluoropyruvate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Methyl 3,3,3-trifluoropyruvate, Methyl 3,3,3-trifluoro-2-oxopropanoate

T1785-1G T1785-5G
TCIAT1785-1GEA 215 SEK
TCIAT1785-1G TCIAT1785-5G
Methyl trifluoropyruvate ≥98.0%
Methyl trifluoropyruvate
Formel: C₄H₃F₃O₃
MW: 156,06 g/mol
Kokpunkt: 86°C
Flampunkt: 26°C
Storage Temperature: Kylskåp
MDL Number: MFCD00114936
CAS nummer: 13089-11-7
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6e49ccb5083916e3d68d67b9f97df7db1fe91a5&vid=e6e49ccb5083916e3d68d67b9f97df7db1fe91a5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR