8-Bromo-2-methylquinoline ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
B3444-1G
B3444-5G
TCIAB3444-1GEA
910
SEK
TCIAB3444-1G
TCIAB3444-5G
8-Bromo-2-methylquinoline ≥98.0% (by GC, titration analysis)
8-Bromo-2-methylquinoline
Formel:
C₁₀H₈BrN Kokpunkt: 132°C |
MDL Number:
MFCD04966997 CAS nummer: 61047-43-6 |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...