2-sec-Butylthiazole ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A13483.06 A13483.14
A13483.06EA 1375 SEK
A13483.06 A13483.14
2-sec-Butylthiazole ≥98%
2-sec-Butylthiazole
Purity: 98+% CAS No.: 18277-27-5 Molecular Formula: C7H11NS Formula Weight: 141.24 Flash Point: 58°(136°F) Refractive Index: 1.4985 Odour: Characteristic Boiling Point: 174-175° UN No.: 1993 MDL No.: MFCD00051951
Formel: C₇H₁₁NS
MW: 141.24 g/mol
Kokpunkt: 174…175 °C
Densitet: 1.007
Flampunkt: 58 °C (136 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00051951
CAS nummer: 18277-27-5
EINECS: 242-154-3
ADR: 3 III