5-(3-Methylphenyl)-1H-tetrazole ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-(m-Tolyl)-1H-tetrazole
L19263.03 L19263.MD
L19263.03EA 4350 SEK
L19263.03 L19263.MD
5-(3-Methylphenyl)-1H-tetrazole ≥99%
5-(3-Methylphenyl)-1H-tetrazole
Purity: 99% CAS No.: 3441-00-7 Molecular Formula: C8H8N4 Formula Weight: 160.18 Melting Point: 143-146° Beilstein Reference: 131205
Formel: C₈H₈N₄
MW: 160,18 g/mol
Smältpunkt: 143…146 °C
Storage Temperature: Omgivande
MDL Number: MFCD03093096
CAS nummer: 3441-00-7