5-(2-Methylphenyl)-1H-tetrazole ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-(o-Tolyl)tetrazole
L19262.03 L19262.MD
L19262.03EA 4025 SEK
L19262.03 L19262.MD
5-(2-Methylphenyl)-1H-tetrazole ≥99%
5-(2-Methylphenyl)-1H-tetrazole
Purity: 99% CAS No.: 51449-86-6 Molecular Formula: C8H8N4 Formula Weight: 160.18 Melting Point: 151-154° Beilstein Reference: 136151
Formel: C₈H₈N₄
MW: 160,18 g/mol
Smältpunkt: 151…154 °C
Storage Temperature: Omgivande
MDL Number: MFCD03093095
CAS nummer: 51449-86-6