Methyl-4-tert-butylbenzoate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Methyl 4-(tert-butyl)benzoate
M1693-25G M1693-500G
TCIAM1693-25GEA 230 SEK
TCIAM1693-25G TCIAM1693-500G
Methyl-4-tert-butylbenzoate ≥98.0%
Methyl-4-tert-butylbenzoate
Formel: C₁₂H₁₆O₂
MW: 192,26 g/mol
Kokpunkt: 247°C
MDL Number: MFCD00008835
CAS nummer: 26537-19-9
EINECS: 247-768-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20e45d3e3028c063468cc183dc4d5b0d6f0cd140&vid=20e45d3e3028c063468cc183dc4d5b0d6f0cd140&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %