3-Phenylthiophene ≥95.0%
Leverantör: TCI EUROPE
P1126-1G
P1126-5G
TCIAP1126-1GEA
1525
SEK
TCIAP1126-1G
TCIAP1126-5G
3-Phenylthiophene ≥95.0%
3-Phenylthiophene
Formel:
C₁₀H₈S MW: 160,235 g/mol Kokpunkt: 256°C |
MDL Number:
MFCD00114997 CAS nummer: 2404-87-7 EINECS: 219-297-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 90.0 to 93.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...