3-(Chloromethyl)-5-(4-methoxyphenyl)-1,2,4-oxadiazole

Leverantör: Thermo Scientific

SEW02033DA SEW02033EA
MAYBSEW02033DAEA 0 SEK
MAYBSEW02033DA MAYBSEW02033EA
3-(Chloromethyl)-5-(4-methoxyphenyl)-1,2,4-oxadiazole
3-(Chloromethyl)-5-(4-methoxyphenyl)-1,2,4-oxadiazole
Formel: C₁₀H₉ClN₂O₂
MDL Number: MFCD00084967
CAS nummer: 73217-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR