3-(Chloromethyl)-5-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole
Leverantör: Thermo Scientific
SEW02034DA
SEW02034EA
MAYBSEW02034DAEA
0
SEK
MAYBSEW02034DA
MAYBSEW02034EA
3-(Chloromethyl)-5-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole
3-(Chloromethyl)-5-(3-(trifluoromethyl)phenyl)-1,2,4-oxadiazole
Formel:
C₁₀H₆ClF₃N₂O |
MDL Number:
MFCD00084969 CAS nummer: 175205-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...