3,5-Bis(trifluoromethyl)thiobenzamide ≥97%
Leverantör: Thermo Scientific
Varning
H26809.MD H26809.03
H26809.MDEA 1600 SEK
H26809.MD H26809.03
3,5-Bis(trifluoromethyl)thiobenzamide ≥97%
3,5-Bis(trifluoromethyl)thiobenzamide
Purity: 97% CAS No.: 317319-15-6 Molecular Formula: C9H5F6NS Formula Weight: 273.20 Melting Point: 112-115° MDL No.: MFCD00278692
Formel: C₉H₅F₆NS
MW: 273,20 g/mol
Smältpunkt: 112…115 °C
Storage Temperature: Omgivande
MDL Number: MFCD00278692
CAS nummer: 317319-15-6