Methyl-3-cyano-4-methoxybenzoate

Leverantör: Thermo Scientific

Synonyms: Methyl 3-cyano-4-methoxybenzoate

TL00306DA TL00306EA
MAYBTL00306DAEA 0 SEK
MAYBTL00306DA MAYBTL00306EA
Methyl-3-cyano-4-methoxybenzoate
Methyl-3-cyano-4-methoxybenzoate
Formel: C₁₀H₉NO₃
MW: 191,19 g/mol
MDL Number: MFCD00052930
CAS nummer: 25978-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR