Methyl-3-cyano-4-methoxybenzoate ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Methyl 3-cyano-4-methoxybenzoate
L11107.03 L11107.06
L11107.03EA 1325 SEK
L11107.03 L11107.06
Methyl-3-cyano-4-methoxybenzoate ≥97%
Methyl-3-cyano-4-methoxybenzoate
Purity: 97% CAS No.: 25978-74-9 Molecular Formula: C10H9NO3 Formula Weight: 191.19 Melting Point: 123-127° Beilstein Reference: 2374769 UN No.: 3439 MDL No.: MFCD00052930
Formel: C₁₀H₉NO₃
MW: 191,19 g/mol
Smältpunkt: 123…127 °C
Storage Temperature: Omgivande
MDL Number: MFCD00052930
CAS nummer: 25978-74-9
ADR: 6.1,III