4-Cyanobenzamide ≥97.0%
Leverantör: TCI
|
Varning
|
C1594-1G
C1594-5G
TCIAC1594-1GEA
750
SEK
TCIAC1594-1G
TCIAC1594-5G
4-Cyanobenzamide ≥97.0%
4-Cyanobenzamide
Formel:
C₈H₆N₂O |
MDL Number:
MFCD00017133 CAS nummer: 3034-34-2 EINECS: 221-223-1 |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 224.0 to 228.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...