α-Bromo-2-chlorophenylacetic acid ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Fara

Synonyms: 2-Bromo-2-(2-chlorophenyl)acetic acid

B2950-25G
TCIAB2950-25GEA 285 SEK
TCIAB2950-25G
α-Bromo-2-chlorophenylacetic acid ≥98.0% (by GC, titration analysis)
α-Bromo-2-chlorophenylacetic acid
Formel: C₈H₆BrClO₂
MW: 249.49 g/mol
MDL Number: MFCD03428513
CAS nummer: 29270-30-2
UN: 3261
ADR: 8 II

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR