1H-Indazole-7-carboxylic acid 95%
Leverantör: Thermo Scientific
Varning
H32463.MD H32463.03
H32463.MDEA 3275 SEK
H32463.MD H32463.03
1H-Indazole-7-carboxylic acid 95%
1H-Indazole-7-carboxylic acid
Purity: 95% CAS No.: 677304-69-7 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: ca 240° dec. MDL No.: MFCD06804572
Formel: C₈H₆N₂O₂
MW: 162,15 g/mol
Smältpunkt: ∼ 240 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD06804572
CAS nummer: 677304-69-7