2-(2-Hydroxyphenyl)benzoxazole ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2-(Benzo[d]oxazol-2-yl)phenol

H0291-1G H0291-25G
TCIAH0291-1GEA 245 SEK
TCIAH0291-1G TCIAH0291-25G
2-(2-Hydroxyphenyl)benzoxazole ≥98.0% (by GC, titration analysis)
2-(2-Hydroxyphenyl)benzoxazole
Formel: C₁₃H₉NO₂
MW: 211,2200 g/mol
Kokpunkt: 338°C
MDL Number: MFCD00005767
CAS nummer: 835-64-3
EINECS: 212-642-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=266f0ca12dc0e560d64629d04dd807b90abfcff3&vid=266f0ca12dc0e560d64629d04dd807b90abfcff3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 123.0 to 126.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR