Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate
A11534.06 A11534.14
A11534.06EA 1625 SEK
A11534.06 A11534.14
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate 98%
Ethyl-5-amino-1-methyl-1H-pyrazole-4-carboxylate
Purity: 98% CAS No.: 31037-02-2 Molecular Formula: C7H11N3O2 Formula Weight: 169.18 Melting Point: 97-102° Beilstein Reference: 131200
Formel: C₇H₁₁N₃O₂
MW: 169,18 g/mol
Smältpunkt: 97…102 °C
Storage Temperature: Omgivande
MDL Number: MFCD00051652
CAS nummer: 31037-02-2