3-Ethynylthiophene ≥97.0%

Leverantör: TCI EUROPE
Varning

E0892-5G E0892-1G
TCIAE0892-5GEA 2250 SEK
TCIAE0892-5G TCIAE0892-1G
3-Ethynylthiophene ≥97.0%
3-Ethynylthiophene
Formel: C₆H₄S
MW: 108,16 g/mol
Kokpunkt: 50°C
Flampunkt: 46°C
Storage Temperature: Kylskåp
MDL Number: MFCD04039973
CAS nummer: 67237-53-0
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa2663a3b3c0fa1c8c226470f148782a4b6856df&vid=fa2663a3b3c0fa1c8c226470f148782a4b6856df&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR