Tri(2-furyl)phosphine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Tri(furan-2-yl)phosphine, Tri-2-furylphosphine

T1643-1G T1643-5G
TCIAT1643-1GEA 615 SEK
TCIAT1643-1G TCIAT1643-5G
Tri(2-furyl)phosphine ≥98.0%
Tri(2-furyl)phosphine
Formel: C₁₂H₉O₃P
MW: 232,17 g/mol
Kokpunkt: 136°C
MDL Number: MFCD00151857
CAS nummer: 5518-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a59699b7d0a1269f1ea8739848a3e0d6422bd900&vid=a59699b7d0a1269f1ea8739848a3e0d6422bd900&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR