6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
Leverantör: Thermo Scientific
BTB07054DA
BTB07054EA
BTB07054EE
MAYBBTB07054DAEA
0
SEK
MAYBBTB07054DA
MAYBBTB07054EA
MAYBBTB07054EE
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
Formel:
C₅H₆N₂O₃ MW: 142,12 g/mol |
MDL Number:
MFCD00052030 CAS nummer: 27372-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...