1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers) ≥97%
Leverantör: Thermo Fisher Scientific
Fara
L20001.22 L20001.36
L20001.22EA 515 SEK
L20001.22 L20001.36
1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers) ≥97%
1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers)
Purity: 97% CAS No.: 85-42-7 Molecular Formula: C8H10O3 Formula Weight: 154.17 Flash Point: 275°(527°F) Melting Point: 32-34° Boiling Point: 158°/17mm Beilstein Reference: 83213 MDL No.: MFCD00064863
Formel: C₈H₁₀O₃
Kokpunkt: 158 °C (17 mmHg)
Smältpunkt: 32…34 °C
Flampunkt: 275 °C (527 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00064863
CAS nummer: 85-42-7
EINECS: 201-604-9